Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 8 | 806 | ||
AK019171.1 | - | 8 | 821 | |||
AK019656.1 | - | 8 | 857 | |||
AK037465.1 | - | 221 | ||||
AK090162.1 | - | 8 | 844 | |||
AK145596.1 | - | 8 | 783 | |||
AK147057.1 | - | 8 | 766 | |||
AK151116.1 | - | 8 | 814 | |||
AK158987.1 | - | 8 | 823 | |||
AK159296.1 | - | 8 | 821 | |||
AK159778.1 | - | 8 | 815 | |||
AK159949.1 | - | 8 | 863 | |||
AK217419.1 | - | 1 | 62 | |||
BC003282.1 | - | 8 | 785 | 3582133 |