Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020600.4 | NP_065625.2 | 3 | 283 | ||
GenBank mRNA | AK003148.1 | - | 3 | 228 | ||
AK011213.1 | - | 3 | 228 | |||
AK012425.1 | - | 3 | 227 | |||
AK012584.1 | - | 3 | 227 | |||
AK019233.1 | - | 3 | 226 | |||
AK028106.1 | - | 3 | 231 | |||
AK146325.1 | - | 3 | 224 | |||
AK146976.1 | - | 3 | 227 | |||
AK152461.1 | - | 1 | 2324 | |||
AK153401.1 | - | 1 | 2324 | |||
AK168237.1 | - | 3 | 227 | |||
BC042940.1 | - | 2 | 207 | 6487681 | ||
BC062874.1 | - | 2 | 206 | 5683900 | ||
BC081449.1 | - | 2 | 194 | 5684375 | ||
BY149058.1 | - | 3 | 284 |