Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IL6R SIRNA Assays ›
          Gene Symbol:
          IL6R
          Gene Name:
          interleukin 6 receptor
          Gene Aliases:
          CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
          Chromosome Location:
          Chr.1: 154405193 - 154469450 on Build GRCh38
          UniGene ID:
          Hs.135087
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7316

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IL6R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000565.3 NP_000556.1 5 1193
          NM_001206866.1 NP_001193795.1 5 1193
          NM_181359.2 NP_852004.1 5 1193
          XM_005245139.1 XP_005245196.1 5 1152
          XM_005245140.2 XP_005245197.1 5 1152
          XM_006711298.1 XP_006711361.1 5 1152
          XM_006711299.3 XP_006711362.1 5 1152
          XM_017001199.1 XP_016856688.1 5 1152
          XM_017001200.1 XP_016856689.1 5 1152
          XM_017001201.1 XP_016856690.1 5 1152
          GenBank mRNA AK223582.1 - 5 1014
          AK293013.1 - 5 807
          AK312730.1 - 5 804
          BC035327.1 - 5 768
          BC089410.1 - 5 1163 3078506
          BC132684.1 - 5 1006 40146706
          BC132686.1 - 5 1006 40146708
          X12830.1 - 5 1193
          X58298.1 - 5 807

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          hepatic immune response
          monocyte chemotaxis
          positive regulation of leukocyte chemotaxis
          acute-phase response
          positive regulation of cell proliferation
          cytokine-mediated signaling pathway
          endocrine pancreas development
          positive regulation of chemokine production
          positive regulation of interleukin-6 production
          response to cytokine
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of osteoblast differentiation
          positive regulation of smooth muscle cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          defense response to Gram-negative bacterium
          interleukin-6-mediated signaling pathway
          ciliary neurotrophic factor-mediated signaling pathway
          extrinsic apoptotic signaling pathway
          ciliary neurotrophic factor receptor activity
          interleukin-6 receptor activity
          interleukin-6 receptor binding
          protein binding
          enzyme binding
          interleukin-6 binding
          protein homodimerization activity
          ciliary neurotrophic factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0