Search
Search

KIF5B| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004521.2 | NP_004512.1 | 12 | 1689 | ||
| XM_017016224.1 | XP_016871713.1 | 12 | 1295 | |||
| GenBank mRNA | AK289413.1 | - | 12 | 1635 | ||
| AK299542.1 | - | 2418 | ||||
| AK307502.1 | - | 13 | 1386 | |||
| BC126279.1 | - | 12 | 1442 | 8991995 | ||
| BC126281.1 | - | 12 | 1442 | 8991997 | ||
| FJ224324.1 | - | 12 | 1532 | |||
| X65873.1 | - | 12 | 1532 | |||