Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Apcs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011318.2 | NP_035448.2 | 2 | 408 | ||
GenBank mRNA | AK144248.1 | - | 2 | 295 | ||
AK165699.1 | - | 2 | 295 | |||
BC061125.1 | - | 2 | 305 | 30308397 | ||
M23552.1 | - | 2 | 384 | |||
X14079.1 | - | 2 | 384 | |||
Y00426.1 | - | 2 | 397 |