Search
Search

Slc20a1| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159593.1 | NP_001153065.1 | 2 | 767 | ||
| NM_015747.2 | NP_056562.1 | 2 | 767 | |||
| GenBank mRNA | AK035898.1 | - | 2 | 738 | ||
| AK084159.1 | - | 2 | 740 | |||
| AK089184.1 | - | 2 | 738 | |||
| AK155506.1 | - | 2 | 740 | |||
| BC015085.1 | - | 2 | 667 | 4015575 | ||
| M73696.1 | - | 1 | 686 | |||