Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC22A1 SIRNA Assays ›
Gene Symbol:
Slc22a1
Gene Name:
solute carrier family 22 (organic cation transporter), member 1
Gene Aliases:
Lx1, Oct1, Orct, Orct1
Chromosome Location:
Chr.17: 12648874 - 12675838 on Build GRCm38
UniGene ID:
Mm.594
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s73893
Size
Availability
Made To Order
Catalog #
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Slc22a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009202.5
NP_033228.2
2
623
GenBank mRNA
AF010259.1
-
2
481
BC021651.1
-
2
623
4211979
U38652.1
-
2
478
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_012697, X78855.
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
ion transport
cation transport
neurotransmitter transport
drug transmembrane transport
establishment or maintenance of transmembrane electrochemical gradient
organic cation transport
quaternary ammonium group transport
monoamine transport
dopamine transport
norepinephrine transport
ion transmembrane transport
epinephrine transport
protein homooligomerization
transmembrane transport
ammonium transmembrane transport
cation transmembrane transport
acetate ester transport
transporter activity
acetylcholine transmembrane transporter activity
dopamine transmembrane transporter activity
norepinephrine transmembrane transporter activity
monoamine transmembrane transporter activity
secondary active organic cation transmembrane transporter activity
ion transmembrane transporter activity
organic cation transmembrane transporter activity
quaternary ammonium group transmembrane transporter activity
transmembrane transporter activity
identical protein binding
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fdqs5:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0