Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGK1 SIRNA Assays ›
          Gene Symbol:
          SGK1
          Gene Name:
          serum/glucocorticoid regulated kinase 1
          Gene Aliases:
          SGK
          Chromosome Location:
          Chr.6: 134169246 - 134318112 on Build GRCh38
          UniGene ID:
          Hs.510078
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s741

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001143676.1 NP_001137148.1 12 1796
          NM_001143677.1 NP_001137149.1 10 1047
          NM_001143678.1 NP_001137150.1 10 1226
          NM_001291995.1 NP_001278924.1 9 870
          NM_005627.3 NP_005618.2 10 1002
          XM_011536071.1 XP_011534373.1 13 1578
          GenBank mRNA AF153609.1 - 10 955
          AK055077.1 - 12 1796
          AK098509.1 - 10 970
          AK293383.1 - 10 1036
          AK295422.1 - 9 838
          AK298688.1 - 10 1285
          BC001263.1 - 10 955 3459056
          BX649005.1 - 2 3575
          EU518415.1 - 12 1198
          FM205707.1 - 10 990
          FM205708.1 - 10 1025
          FM205709.1 - 12 1645
          FM205710.1 - 9 1114
          Y10032.1 - 10 955

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          protein phosphorylation
          sodium ion transport
          cellular sodium ion homeostasis
          apoptotic process
          response to stress
          cellular response to DNA damage stimulus
          microtubule depolymerization
          long-term memory
          regulation of blood pressure
          visual learning
          positive regulation of sodium ion transport
          peptidyl-serine phosphorylation
          positive regulation of cell growth
          regulation of cell migration
          negative regulation of microtubule polymerization
          positive regulation of transporter activity
          ion transmembrane transport
          intracellular signal transduction
          regulation of cell proliferation
          regulation of apoptotic process
          negative regulation of apoptotic process
          glucocorticoid mediated signaling pathway
          neuron projection morphogenesis
          positive regulation of dendrite morphogenesis
          regulation of catalytic activity
          regulation of sequence-specific DNA binding transcription factor activity
          regulation of cell cycle
          regulation of gastric acid secretion
          renal sodium ion absorption
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          calcium channel regulator activity
          protein binding
          ATP binding
          potassium channel regulator activity
          sodium channel regulator activity
          chloride channel regulator activity
          3-phosphoinositide-dependent protein kinase binding
          cofactor binding
          tau protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0