Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 11 | 1177 | ||
NM_001102605.1 | NP_001096075.1 | 10 | 1147 | |||
NM_001142573.1 | NP_001136045.1 | 8 | 947 | |||
NM_001142574.1 | NP_001136046.1 | 8 | 932 | |||
NM_001142575.1 | NP_001136047.1 | 7 | 872 | |||
NM_001142576.1 | NP_001136048.1 | 10 | 1078 | |||
NM_001304521.1 | NP_001291450.1 | 8 | 975 | |||
NM_183243.2 | NP_899066.1 | 9 | 1069 | |||
XM_017012168.1 | XP_016867657.1 | 10 | 1147 | |||
XM_017012169.1 | XP_016867658.1 | 9 | 1069 | |||
XM_017012170.1 | XP_016867659.1 | 8 | 976 | |||
XM_017012171.1 | XP_016867660.1 | 10 | 1265 | |||
XM_017012172.1 | XP_016867661.1 | 10 | 1265 | |||
XM_017012173.1 | XP_016867662.1 | 10 | 1147 | |||
GenBank mRNA | AK054640.1 | - | 7 | 844 | ||
AK054667.1 | - | 9 | 1069 | |||
AK092452.1 | - | 10 | 1114 | |||
AK122994.1 | - | 10 | 1146 | |||
AK131294.1 | - | 8 | 894 | |||
AK293413.1 | - | 8 | 904 | |||
AK297943.1 | - | 8 | 919 | |||
BC033622.1 | - | 9 | 1060 | 5210847 | ||
CD014008.1 | - | 9 | 861 | |||
CD014010.1 | - | 10 | 983 | |||
CR933672.1 | - | 1113 | ||||
J05272.1 | - | 11 | 1441 |