Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sprr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204427.1 | NP_001191356.1 | 2 | 660 | ||
NM_011478.2 | NP_035608.1 | 2 | 796 | |||
GenBank mRNA | BC125629.1 | - | 1 | 690 | 40130144 |