Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRF SIRNA Assays ›
Gene Symbol:
Srf
Gene Name:
serum response factor
Gene Aliases:
AW049942, AW240594
Chromosome Location:
Chr.17: 46546838 - 46556870 on Build GRCm38
UniGene ID:
Mm.45044
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s74391
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Srf
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_020493.2
NP_065239.1
1
770
GenBank mRNA
BC051950.1
-
1
783
6404515
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_003131, BC048211, BC052572, AB209128, J03161.
This siRNA also targets the following transcripts in Rat: NM_001109302, XM_006244557, XM_006244558.
Gene Ontology Categories:
Function(s)
Process(es)
patterning of blood vessels
in utero embryonic development
mesoderm formation
neuron migration
trophectodermal cell differentiation
heart looping
morphogenesis of an epithelial sheet
cell migration involved in sprouting angiogenesis
leukocyte differentiation
positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
actin filament organization
cell-matrix adhesion
multicellular organism development
gastrulation
heart development
long-term memory
negative regulation of cell proliferation
associative learning
response to hormone
epithelial structure maintenance
positive regulation of transcription via serum response element binding
single organismal cell-cell adhesion
hippocampus development
tangential migration from the subventricular zone to the olfactory bulb
actin cytoskeleton organization
contractile actin filament bundle assembly
regulation of cell adhesion
platelet activation
platelet formation
negative regulation of cell migration
neuron projection development
regulation of water loss via skin
response to cytokine
megakaryocyte development
dorsal aorta morphogenesis
mRNA transcription from RNA polymerase II promoter
stress fiber assembly
skin morphogenesis
positive thymic T cell selection
sarcomere organization
positive regulation of cell differentiation
positive regulation of axon extension
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle contraction
positive regulation of transcription by glucose
muscle cell cellular homeostasis
developmental growth
erythrocyte development
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of filopodium assembly
cardiac myofibril assembly
hematopoietic stem cell differentiation
positive regulation of transcription initiation from RNA polymerase II promoter
long term synaptic depression
heart trabecula formation
lung morphogenesis
bronchus cartilage development
trachea cartilage development
cardiac vascular smooth muscle cell differentiation
eyelid development in camera-type eye
lung smooth muscle development
bicellular tight junction assembly
primitive streak formation
epithelial cell-cell adhesion
cellular senescence
negative regulation of beta-amyloid clearance
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter sequence-specific
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
transcription factor binding
serum response element binding
chromatin DNA binding
protein homodimerization activity
histone deacetylase binding
sequence-specific DNA binding
protein dimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0