Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AURKA SIRNA Assays ›
          Gene Symbol:
          Aurka
          Gene Name:
          aurora kinase A
          Gene Aliases:
          AIRK1, ARK-1, AU019385, AW539821, Ark1, Aurora-A, Ayk1, IAK, IAK1, Stk6
          Chromosome Location:
          Chr.2: 172356190 - 172370706 on Build GRCm38
          UniGene ID:
          Mm.249363
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s74515

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Aurka
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291185.1 NP_001278114.1 8, 9 1103
          NM_011497.4 NP_035627.1 8, 9 1181
          GenBank mRNA AF007817.1 - 8, 9 1128
          AK077513.1 - 8, 9 1059
          AK085861.1 - 8, 9 1055
          AK145968.1 - 8, 9 1069
          AK169363.1 - 8, 9 1058
          AK189850.1 - 1, 2 30
          AK213221.1 - 1, 2 32
          AY336976.1 - 7, 8 1005
          BC005425.1 - 8, 9 1048 3590706
          BC014711.1 - 8, 9 1117 3968835
          U69106.1 - 8, 9 1039
          U80932.1 - 8, 9 1048

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic spindle organization
          microtubule cytoskeleton organization
          mitotic cell cycle
          protein phosphorylation
          cell cycle
          spindle organization
          mitotic spindle organization
          spindle assembly involved in female meiosis I
          mitotic nuclear division
          mitotic centrosome separation
          meiotic nuclear division
          anterior/posterior axis specification
          phosphorylation
          cell projection organization
          regulation of protein stability
          negative regulation of protein binding
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of cytokinesis
          histone-serine phosphorylation
          negative regulation of apoptotic process
          centrosome organization
          cell division
          centrosome localization
          protein localization to centrosome
          negative regulation of spindle checkpoint
          positive regulation of oocyte maturation
          neuron projection extension
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          ubiquitin protein ligase binding
          histone serine kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qq54g:80/100.66.72.153:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0