Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TAP2 SIRNA Assays ›
          Gene Symbol:
          Tap2
          Gene Name:
          transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
          Gene Aliases:
          ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas
          Chromosome Location:
          Chr.17: 34204085 - 34216321 on Build GRCm38
          UniGene ID:
          Mm.14814
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s74736

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tap2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011530.3 NP_035660.3 12 2153
          GenBank mRNA AK020022.1 - 11 2229
          AK079667.1 - 12 2067
          AK145234.1 - 12 2112
          AK151376.1 - 12 2063
          AK151483.1 - 12 2063
          AK151657.1 - 12 2086
          AK153287.1 - 12 2063
          AK159200.1 - 12 2119
          AK161091.1 - 12 2069
          AK169645.1 - 9 1361
          BC005578.1 - 12 2053 3968225
          BC051257.1 - 12 2066 6530488
          BC057668.1 - 12 2440
          CT010387.1 - 11 2000
          KJ650229.1 - 11 2003
          M90459.1 - 11 2000
          U60087.1 - 11 2003
          U60088.1 - 11 2003
          U60089.1 - 11 2003
          U60090.1 - 11 2003
          U60091.1 - 11 2003
          U60092.1 - 11 2000

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_032056, XM_008772777, KC222910, X75307, X63854, and others.

          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          immune system process
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
          antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
          positive regulation of antigen processing and presentation of peptide antigen via MHC class I
          protein complex assembly
          transport
          protein transport
          peptide transport
          intracellular transport of viral protein in host cell
          antigen processing and presentation of endogenous peptide antigen via MHC class I
          protection from natural killer cell mediated cytotoxicity
          cytosol to ER transport
          peptide antigen transport
          transmembrane transport
          nucleotide binding
          protein binding
          ATP binding
          peptide transporter activity
          peptide antigen-transporting ATPase activity
          peptide-transporting ATPase activity
          ATPase activity
          MHC class Ib protein binding
          MHC protein binding
          MHC class I protein binding
          peptide antigen binding
          ATPase activity, coupled to transmembrane movement of substances
          protein homodimerization activity
          ADP binding
          TAP1 binding
          TAP2 binding
          tapasin binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gwjk7:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0