Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tfec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031198.3 | NP_112475.1 | 7 | 993 | ||
GenBank mRNA | AF077742.1 | - | 7 | 993 | ||
AK137141.1 | - | 7 | 958 | |||
AK156327.1 | - | 7 | 906 | |||
AK159579.1 | - | 7 | 926 | |||
BC052832.1 | - | 6 | 881 | |||
BC064770.1 | - | 6 | 869 | |||
BC098494.1 | - | 7 | 878 | 6431607 |