Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGA6 SIRNA Assays ›
          Gene Symbol:
          ITGA6
          Gene Name:
          integrin subunit alpha 6
          Gene Aliases:
          CD49f, ITGA6B, VLA-6
          Chromosome Location:
          Chr.2: 172427354 - 172506455 on Build GRCh38
          UniGene ID:
          Hs.133397
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7492
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITGA6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000210.3 NP_000201.2 2 529
          NM_001079818.2 NP_001073286.1 2 529
          NM_001316306.1 NP_001303235.1 2 230
          XM_006712510.1 XP_006712573.1 2 498
          XM_006712511.1 XP_006712574.1 2 498
          XM_017004005.1 XP_016859494.1 2 225
          XM_017004006.1 XP_016859495.1 2 225
          XM_017004007.1 XP_016859496.1 2 225
          XM_017004008.1 XP_016859497.1 2 225
          GenBank mRNA AB208842.1 - 2 197
          AK296496.1 - 2 166
          AK311695.1 - 2 230
          BC007002.1 - 2 495
          BC136455.1 - 2 403 9020444
          BC136456.1 - 2 403 9020445
          DA760250.1 - 2 498
          X53586.1 - 2 441
          X59512.1 - 2 277

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00173952_m1

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-substrate junction assembly
          cell-matrix adhesion
          integrin-mediated signaling pathway
          ectodermal cell differentiation
          positive regulation of cell-cell adhesion
          extracellular matrix organization
          hemidesmosome assembly
          cell-substrate adhesion
          cellular response to extracellular stimulus
          cell adhesion mediated by integrin
          nail development
          positive regulation of phosphorylation
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          skin development
          positive regulation of transcription from RNA polymerase II promoter
          filopodium assembly
          digestive tract development
          brown fat cell differentiation
          leukocyte migration
          renal system development
          amelogenesis
          cell-cell adhesion
          negative regulation of extrinsic apoptotic signaling pathway
          integrin binding
          protein binding
          insulin-like growth factor I binding
          neuregulin binding
          metal ion binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0