Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGA5 SIRNA Assays ›
          Gene Symbol:
          ITGA5
          Gene Name:
          integrin subunit alpha 5
          Gene Aliases:
          CD49e, FNRA, VLA-5, VLA5A
          Chromosome Location:
          Chr.12: 54395261 - 54419266 on Build GRCh38
          UniGene ID:
          Hs.505654
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s7548
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ITGA5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002205.4 NP_002196.4 24 2480
          GenBank mRNA AK291570.1 - 24 2425
          AK312414.1 - 24 2480
          BC008786.2 - 24 2465 3629647
          BX647780.1 - 13 2223
          M13918.2 - 24 2367
          X06256.1 - 24 2435

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00233732_m1

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          cell-substrate junction assembly
          cell adhesion
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          leukocyte cell-cell adhesion
          integrin-mediated signaling pathway
          memory
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          positive regulation of cell migration
          positive regulation of vascular endothelial growth factor receptor signaling pathway
          cell-substrate adhesion
          cell-cell adhesion mediated by integrin
          heterotypic cell-cell adhesion
          wound healing, spreading of epidermal cells
          endodermal cell differentiation
          viral entry into host cell
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          positive regulation of sprouting angiogenesis
          negative regulation of anoikis
          virus receptor activity
          epidermal growth factor receptor binding
          platelet-derived growth factor receptor binding
          integrin binding
          protein binding
          vascular endothelial growth factor receptor 2 binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0