Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009485.1 | NP_033511.1 | 4 | 1526 | ||
GenBank mRNA | AF011411.1 | - | 4 | 1526 |
Set Membership: |
G-protein coupled receptor |