Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other VMN2R37 SIRNA Assays ›
          Gene Symbol:
          Vmn2r37 Vmn2r30 Vmn2r32 Vmn2r34 Vmn2r36 Vmn2r40 Vmn2r41 Vmn2r45 Vmn2r47 Vmn2r48 Vmn2r49 Vmn2r51
          Gene Name:
          vomeronasal 2, receptor 37
          vomeronasal 2, receptor 30
          vomeronasal 2, receptor 32
          vomeronasal 2, receptor 34
          vomeronasal 2, receptor 36
          vomeronasal 2, receptor 40
          vomeronasal 2, receptor 41
          vomeronasal 2, receptor 45
          vomeronasal 2, receptor 47
          vomeronasal 2, receptor 48
          vomeronasal 2, receptor 49
          vomeronasal 2, receptor 51
          Gene Aliases:
          V2r14
          ENSMUSG00000070847, V2r15
          V2r5
          ENSMUSG00000070841
          100042724, Gm3991
          -
          -
          -
          -
          EG625580
          -
          ENSMUSG00000058685
          Chromosome Location:
          Chr.7: 9205549 - 9223653 on Build GRCm38
          Chromosome Location:
          Chr.7: 7312274 - 7337493 on Build GRCm38
          Chromosome Location:
          Chr.7: 7463969 - 7479973 on Build GRCm38
          Chromosome Location:
          Chr.7: 7671829 - 7689398 on Build GRCm38
          Chromosome Location:
          Chr.7: 7876653 - 7902462 on Build GRCm38
          Chromosome Location:
          Chr.7: 8907734 - 8931402 on Build GRCm38
          Chromosome Location:
          Chr.7: 8137905 - 8161551 on Build GRCm38
          Chromosome Location:
          Chr.7: 8471469 - 8488959 on Build GRCm38
          Chromosome Location:
          Chr.7: 9816782 - 9841325 on Build GRCm38
          Chromosome Location:
          Chr.7: 9927624 - 9953585 on Build GRCm38
          Chromosome Location:
          Chr.7: 9976245 - 9992139 on Build GRCm38
          Chromosome Location:
          Chr.7: 10087198 - 10105659 on Build GRCm38
          UniGene ID:
          Mm.302158
          Mm.458977
          Mm.482571
          Mm.302158
          Mm.465202
          Mm.469893
          Mm.469892
          Mm.469890
          Mm.465205
          Mm.469893
          Mm.484763
          Mm.291126
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s75867

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Vmn2r37
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009489.2 NP_033515.2 1 295
          GenBank mRNA AF011424.1 - 1 295
          Gene Symbol: Vmn2r30
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009490.3 NP_033516.3 1 180
          GenBank mRNA AF011425.1 - 218
          Gene Symbol: Vmn2r32
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105063.1 NP_001098533.1 1 180
          Gene Symbol: Vmn2r34
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105066.1 NP_001098536.1 1 180
          Gene Symbol: Vmn2r36
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105068.1 NP_001098538.1 1 180
          Gene Symbol: Vmn2r40
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105072.1 NP_001098542.1 1 180
          Gene Symbol: Vmn2r41
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105073.1 NP_001098543.1 1 180
          Gene Symbol: Vmn2r45
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105075.1 NP_001098545.1 1 180
          Gene Symbol: Vmn2r47
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105151.1 NP_001098621.1 1 180
          Gene Symbol: Vmn2r48
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105152.1 NP_001098622.1 1 189
          Gene Symbol: Vmn2r49
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105156.1 NP_001098626.1 1 180
          Gene Symbol: Vmn2r51
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105179.1 NP_001098649.1 1 180

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xhq5n:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0