Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vps45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013841.3 | NP_038869.1 | 1, 2 | 781 | ||
GenBank mRNA | AK083678.1 | - | 1, 2 | 157 | ||
AK146190.1 | - | 1, 2 | 183 | |||
AK168101.1 | - | 1, 2 | 183 | |||
BC012691.1 | - | 1, 2 | 157 | 4038142 | ||
BC058528.1 | - | 1, 2 | 173 | 5718352 | ||
U66865.1 | - | 1, 2 | 796 |