Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT3A SIRNA Assays ›
          Gene Symbol:
          Wnt3a
          Gene Name:
          wingless-type MMTV integration site family, member 3A
          Gene Aliases:
          Wnt-3a, vt
          Chromosome Location:
          Chr.11: 59248036 - 59290751 on Build GRCm38
          UniGene ID:
          Mm.1367
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s76081

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Wnt3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009522.2 NP_033548.1 3 459
          GenBank mRNA X56842.1 - 3 483

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          somitogenesis
          positive regulation of cytokine production
          positive regulation of protein phosphorylation
          heart looping
          positive regulation of receptor internalization
          negative regulation of heart induction by canonical Wnt signaling pathway
          signal transduction
          cell-cell signaling
          multicellular organism development
          determination of left/right symmetry
          axonogenesis
          axon guidance
          mesoderm development
          positive regulation of cell proliferation
          organ morphogenesis
          anterior/posterior pattern specification
          COP9 signalosome assembly
          negative regulation of neuron projection development
          Wnt signaling pathway
          spinal cord association neuron differentiation
          hippocampus development
          cell proliferation in forebrain
          Wnt signaling pathway involved in forebrain neuroblast division
          dorsal/ventral neural tube patterning
          hemopoiesis
          platelet activation
          extracellular matrix organization
          BMP signaling pathway
          mammary gland development
          positive regulation of B cell proliferation
          positive regulation of protein binding
          positive regulation of peptidyl-serine phosphorylation
          cellular protein localization
          positive regulation of catenin import into nucleus
          skeletal muscle cell differentiation
          post-anal tail morphogenesis
          inner ear morphogenesis
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          somatic stem cell division
          positive regulation of mesodermal cell fate specification
          paraxial mesodermal cell fate commitment
          positive regulation of collateral sprouting in absence of injury
          negative regulation of axon extension involved in axon guidance
          negative regulation of neurogenesis
          regulation of axonogenesis
          positive regulation of sequence-specific DNA binding transcription factor activity
          palate development
          canonical Wnt signaling pathway
          cardiac muscle cell fate commitment
          positive regulation of protein tyrosine kinase activity
          positive regulation of dermatome development
          canonical Wnt signaling pathway involved in cardiac muscle cell fate commitment
          regulation of microtubule cytoskeleton organization
          platelet aggregation
          axis elongation involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          positive regulation of cell-cell adhesion mediated by cadherin
          positive regulation of canonical Wnt signaling pathway involved in controlling type B pancreatic cell proliferation
          positive regulation of neural precursor cell proliferation
          positive regulation of receptor activity
          positive regulation of cardiac muscle cell differentiation
          transcription coactivator activity
          receptor binding
          frizzled binding
          frizzled-2 binding
          protein binding
          protein domain specific binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0