Search Thermo Fisher Scientific
Gene Symbol: JAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002226.4 | NP_002217.3 | 13 | 2092 | ||
NM_145159.2 | NP_660142.1 | 12 | 1978 | |||
GenBank mRNA | AF003521.1 | - | 13 | 1720 | ||
AF020201.1 | - | 13 | 1739 | |||
AF029778.1 | - | 13 | 2092 | |||
AF029779.1 | - | 12 | 1978 | |||
BC032053.1 | - | 13 | 1756 | |||
U69180.1 | - | 4 | 350 | |||
Y14330.1 | - | 13 | 1643 |