Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145947.1 | NP_001139419.1 | 2 | 786 | ||
NM_011792.5 | NP_035922.4 | 2 | 786 | |||
GenBank mRNA | AF190726.2 | - | 2 | 714 | ||
AF200346.1 | - | 2 | 287 | |||
AK014464.1 | - | 2 | 716 | |||
AK033112.1 | - | 2 | 736 | |||
AK041285.1 | - | 2 | 736 | |||
AK046175.1 | - | 2 | 733 | |||
AK049626.1 | - | 2 | 719 | |||
AK079814.1 | - | 1 | 192 | |||
AK080498.1 | - | 2 | 738 | |||
AK082230.1 | - | 2 | 734 | |||
AK082317.1 | - | 2 | 737 | |||
AK163583.1 | - | 2 | 737 | |||
AK185690.1 | - | 2 | 76 | |||
AK215510.1 | - | 2 | 75 | |||
BC048189.1 | - | 2 | 712 | 6831622 |