Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 12 | 1231 | ||
NM_001253784.1 | NP_001240713.1 | 11 | 1209 | |||
NM_001281803.1 | NP_001268732.1 | 10 | 1213 | |||
NM_054046.5 | NP_473387.1 | 10 | 1122 | |||
GenBank mRNA | AK028726.1 | - | 11 | 1181 | ||
AK034028.1 | - | 1088 | ||||
AK046271.1 | - | 10 | 1082 | |||
AK050405.1 | - | 11 | 1180 | |||
AK081295.1 | - | 10 | 1112 | |||
AK086174.1 | - | 10 | 1085 | |||
AK087778.1 | - | 10 | 1093 | |||
AK129019.1 | - | 10 | 1164 | |||
AK133979.1 | - | 10 | 1096 | |||
AK150334.1 | - | 10 | 1058 | |||
AK150994.1 | - | 10 | 1058 | |||
AK164147.1 | - | 10 | 1094 | |||
BC003306.1 | - | 12 | 1182 | 3154712 | ||
CN705834.1 | - | 3 | 163 | |||
X96708.1 | - | 2 | 91 |