Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DNAJB6 SIRNA Assays ›
          Gene Symbol:
          Dnajb6
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 6
          Gene Aliases:
          Mrj, mDj4
          Chromosome Location:
          Chr.5: 29735687 - 29786478 on Build GRCm38
          UniGene ID:
          Mm.290110
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s76678

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dnajb6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001037940.4 NP_001033029.1 5 581
          NM_001037941.3 NP_001033030.2 5 581
          NM_001127367.1 NP_001120839.1 5 581
          NM_011847.4 NP_035977.2 5 581
          GenBank mRNA AB028854.1 - 5 483
          AF035962.1 - 5 481
          AK005680.1 - 5 486
          AK032039.1 - 5 490
          AK133805.1 - 5 498
          AK134387.1 - 5 540
          AK144182.1 - 5 508
          AK151219.1 - 5 488
          AK151752.1 - 5 455
          AK152626.1 - 5 495
          AK152632.1 - 5 455
          AK153091.1 - 5 461
          AK169332.1 - 5 487
          AK169767.1 - 5 452
          AK180819.1 - 2 155
          AK184807.1 - 1 54
          AK185468.1 - 1 55
          AK213923.1 - 1 54
          AK214355.1 - 1 54
          AK215372.1 - 1 50
          BC003702.1 - 5 463 3601332
          BC083349.1 - 5 473 30541723
          BG800394.1 - 5 471
          DV660970.1 - 4 376

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_058246, NM_005494, CR533498, AK297989, AK297796, and others.
          This siRNA also targets the following transcripts in Rat: NM_001013209, XM_006235863, XM_006235866, XM_001073579, XM_006235864, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          actin cytoskeleton organization
          extracellular matrix organization
          positive regulation of ATPase activity
          protein localization to nucleus
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          intermediate filament organization
          negative regulation of transcription, DNA-templated
          chorio-allantoic fusion
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          chorion development
          chaperone-mediated protein folding
          negative regulation of inclusion body assembly
          ATPase activator activity
          DNA binding
          protein binding
          heat shock protein binding
          protein binding involved in protein folding
          unfolded protein binding
          chaperone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0