Search
Search

KCNJ2| Gene Symbol: KCNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000891.2 | NP_000882.1 | 2 | 1156 | ||
| GenBank mRNA | AF011904.1 | - | 1 | 773 | ||
| AF021139.1 | - | 1 | 773 | |||
| AF153820.1 | - | 2 | 1156 | |||
| AK314081.1 | - | 2 | 1174 | |||
| U12507.1 | - | 2 | 1098 | |||
| U16861.1 | - | 2 | 1100 | |||
| U22413.1 | - | 1 | 773 | |||
| U24055.1 | - | 1 | 886 | |||