Search
Search

Ecsit| Gene Symbol: Ecsit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253897.1 | NP_001240826.1 | 8 | 1280 | ||
| NM_001253898.1 | NP_001240827.1 | 7 | 1102 | |||
| NM_012029.2 | NP_036159.1 | 8 | 1288 | |||
| GenBank mRNA | AF182510.1 | - | 7 | 1301 | ||
| AK007564.1 | - | 8 | 1254 | |||
| AK012835.1 | - | 8 | 1253 | |||
| BC004583.1 | - | 7 | 1089 | 3587327 | ||
| BC016424.1 | - | 8 | 1228 | 4924678 | ||