Search Thermo Fisher Scientific
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033988.2 | NP_001029160.1 | 8 | 1238 | ||
NM_001284319.1 | NP_001271248.1 | 8 | 1230 | |||
NM_019744.4 | NP_062718.2 | 8 | 1148 | |||
GenBank mRNA | AF159461.1 | - | 8 | 1145 | ||
AK014479.1 | - | 8 | 1146 | |||
AK032951.1 | - | 6 | 904 | |||
AK129020.1 | - | 8 | 1196 | |||
AK143965.1 | - | 8 | 1142 | |||
AK145070.1 | - | 8 | 1199 | |||
AK145945.1 | - | 8 | 1191 | |||
AK145954.1 | - | 8 | 1195 | |||
AK146384.1 | - | 8 | 1149 | |||
AK146498.1 | - | 8 | 1188 | |||
AK146598.1 | - | 8 | 1172 | |||
AK146646.1 | - | 8 | 1143 | |||
AK146649.1 | - | 8 | 1145 | |||
AK147036.1 | - | 8 | 1146 | |||
AK147125.1 | - | 8 | 1190 | |||
AK156999.1 | - | 8 | 1230 | |||
BC031528.1 | - | 8 | 1189 | 4008986 | ||
BC085086.1 | - | 8 | 1134 | 6335192 |