Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178600.2 | NP_848715.1 | 1 | 100 | ||
GenBank mRNA | AK002742.1 | - | 1 | 81 | ||
AK003237.1 | - | 1 | 101 | |||
AK008509.1 | - | 1 | 74 | |||
AK008578.1 | - | 1 | 75 | |||
AK013996.1 | - | 1 | 84 | |||
BC031732.1 | - | 1 | 82 | 3991412 |