Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Vkorc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178600.2 | NP_848715.1 | 3 | 471 | ||
GenBank mRNA | AK002742.1 | - | 3 | 452 | ||
AK003237.1 | - | 3 | 472 | |||
AK008509.1 | - | 3 | 445 | |||
AK008578.1 | - | 3 | 446 | |||
AK013996.1 | - | 3 | 455 | |||
BC031732.1 | - | 3 | 453 | 3991412 |