Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199015.1 | NP_001185944.1 | 7, 8 | 912 | ||
NM_001199016.1 | NP_001185945.1 | 6, 7 | 800 | |||
NM_021291.3 | NP_067266.1 | 6, 7 | 1027 | |||
GenBank mRNA | AF192310.1 | - | 7, 8 | 912 | ||
AJ249198.1 | - | 6, 7 | 1027 | |||
AK142541.1 | - | 6, 7 | 801 | |||
AK164714.1 | - | 5, 6 | 1217 | |||
BC010746.1 | - | 7, 8 | 890 | 4236578 |