Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prok2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037539.2 | NP_001032628.1 | 3 | 1272 | ||
NM_001170419.1 | NP_001163890.1 | 2 | 1146 | |||
NM_015768.2 | NP_056583.1 | 4 | 1335 | |||
GenBank mRNA | AF182064.1 | - | 4 | 1333 | ||
AF182066.1 | - | 3 | 1270 | |||
AK015462.1 | - | 4 | 1296 |