Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fbxo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015798.3 | NP_056613.1 | 5 | 637 | ||
GenBank mRNA | AF176530.1 | - | 5 | 621 | ||
AK082809.1 | - | 5 | 658 | |||
AK163224.1 | - | 5 | 638 | |||
AK166591.1 | - | 5 | 638 | |||
AK166599.1 | - | 5 | 639 | |||
AK166918.1 | - | 5 | 634 | |||
AK167118.1 | - | 5 | 638 | |||
AK167270.1 | - | 5 | 637 |