Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pkd2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163004.1 | NP_001156476.1 | 11 | 1608 | ||
NM_016927.3 | NP_058623.2 | 11 | 1608 | |||
GenBank mRNA | AF182033.1 | - | 11 | 1608 | ||
AK030120.1 | - | 1475 | ||||
AK133423.1 | - | 11 | 1605 | |||
BC138717.1 | - | 11 | 1593 | 8861739 |