Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 10 | 2155 | ||
NM_001285871.1 | NP_001272800.1 | 10 | 2340 | |||
NM_001285873.1 | NP_001272802.1 | 10 | 2155 | |||
NM_018880.3 | NP_061368.1 | 10 | 2129 | |||
GenBank mRNA | AB030912.1 | - | 10 | 2156 | ||
AF220019.1 | - | 10 | 2048 | |||
AK019165.1 | - | 10 | 2090 | |||
AK144674.1 | - | 10 | 2058 | |||
AK170029.1 | - | 10 | 2093 | |||
AK182206.1 | - | 1 | 68 | |||
BC003347.1 | - | 10 | 2169 | |||
BC034263.1 | - | 10 | 2094 | 4503884 |