Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019493.3 | NP_062366.2 | 2 | 268 | ||
GenBank mRNA | AB050983.2 | - | 2 | 268 | ||
AJ005120.1 | - | 2 | 141 | |||
AK136041.1 | - | 2 | 255 | |||
AK136124.1 | - | 2 | 256 | |||
AK136149.1 | - | 2 | 258 | |||
AK136185.1 | - | 2 | 260 | |||
AK161379.1 | - | 4 | 347 | |||
AK162122.1 | - | 2 | 255 | |||
AK163407.1 | - | 2 | 254 | |||
BC066811.1 | - | 2 | 242 | 30475849 |