Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039137.3 | NP_001034226.1 | 4 | 360 | ||
NM_001285992.1 | NP_001272921.1 | 4 | 462 | |||
NM_019708.4 | NP_062682.1 | 4 | 465 | |||
GenBank mRNA | AF115778.1 | - | 4 | 283 | ||
AK002521.1 | - | 4 | 314 | |||
AK003095.1 | - | 4 | 308 | |||
AK076085.1 | - | 4 | 258 | |||
AK167488.1 | - | 4 | 308 | |||
AK207171.1 | - | 2 | 141 | |||
BC017629.1 | - | 4 | 284 | |||
BC085265.1 | - | 4 | 272 | 6391098 | ||
BC092071.1 | - | 4 | 268 | 4505111 | ||
BC108407.1 | - | 4 | 282 | |||
BC147069.1 | - | 4 | 353 | 9056075 | ||
CX241959.1 | - | 4 | 451 |