Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LAMA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005560.4 | NP_005551.3 | 36 | 4852 | ||
XM_006723796.3 | XP_006723859.1 | 36 | 4855 | |||
XM_006723798.3 | XP_006723861.1 | 36 | 4858 | |||
XM_011528818.2 | XP_011527120.1 | 35 | 4717 | |||
XM_011528819.2 | XP_011527121.1 | 34 | 4582 | |||
GenBank mRNA | AB010099.1 | - | 2 | 272 | ||
AB067494.1 | - | 33 | 4196 | |||
AF443072.1 | - | 36 | 4852 |