Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: Apba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018758.2 | NP_061228.1 | 5 | 1063 | ||
GenBank mRNA | AF070975.1 | - | 5 | 1017 | ||
AK045965.1 | - | 5 | 1064 | |||
AK082046.1 | - | 983 | ||||
AK169462.1 | - | 5 | 1047 | |||
AK170182.1 | - | 5 | 1011 | |||
BC005423.1 | - | 5 | 995 | 3487448 | ||
BC009666.1 | - | 5 | 1007 | 3490710 |