Search
Search

LDHB| Gene Symbol: LDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174097.2 | NP_001167568.1 | 5 | 776 | ||
| NM_001315537.1 | NP_001302466.1 | 5 | 554 | |||
| NM_002300.7 | NP_002291.1 | 5 | 554 | |||
| XM_006719074.2 | XP_006719137.1 | 5 | 670 | |||
| GenBank mRNA | AK129881.1 | - | 5 | 505 | ||
| AK312328.1 | - | 5 | 538 | |||
| AL516910.3 | - | 5 | 775 | |||
| AY189689.1 | - | 5 | 530 | |||
| BC002362.1 | - | 5 | 521 | 2961445 | ||
| BC015122.1 | - | 5 | 545 | 3685985 | ||
| BC071860.1 | - | 5 | 486 | 4902461 | ||
| BT019764.1 | - | 4 | 443 | |||
| BT019765.1 | - | 4 | 443 | |||
| BU540074.1 | - | 5 | 557 | |||
| CX867875.1 | - | 5 | 751 | |||
| EU794633.1 | - | 5 | 497 | |||
| Y00711.1 | - | 5 | 497 | |||