Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020575.2 | NP_065600.1 | 8, 9 | 2166 | ||
GenBank mRNA | AF155739.1 | - | 8, 9 | 2166 | ||
AK080468.1 | - | 1, 2 | 595 | |||
AK143177.1 | - | 8, 9 | 2188 | |||
AK166666.1 | - | 8, 9 | 2173 | |||
AK167246.1 | - | 8, 9 | 2148 | |||
AK168555.1 | - | 8, 9 | 2169 | |||
BC025029.1 | - | 8, 9 | 2130 | 4935124 |