Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000230.2 | NP_000221.1 | 3 | 343 | ||
XM_005250340.4 | XP_005250397.1 | 3 | 417 | |||
GenBank mRNA | AK313553.1 | - | 3 | 341 | ||
BC060830.1 | - | 3 | 341 | 30333193 | ||
BC069323.1 | - | 2 | 337 | 7262097 | ||
BC069452.1 | - | 2 | 337 | 7262109 | ||
BC069527.1 | - | 2 | 337 | 7262121 | ||
D49487.1 | - | 2 | 300 | |||
U18915.1 | - | 2 | 286 | |||
U43653.1 | - | 2 | 342 |