Search Thermo Fisher Scientific
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021347.4 | NP_067322.1 | 10, 11 | 1124 | ||
GenBank mRNA | AB033595.1 | - | 10, 11 | 1113 | ||
AK028645.1 | - | 10, 11 | 1125 | |||
AK028698.1 | - | 10, 11 | 1125 | |||
AK029062.1 | - | 10, 11 | 1108 |