Search
Search

Smpd3| Gene Symbol: Smpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021491.3 | NP_067466.1 | 5 | 1921 | ||
| GenBank mRNA | AJ250461.1 | - | 3 | 1452 | ||
| AK019476.2 | - | 6 | 1973 | |||
| AK076416.1 | - | 5 | 1898 | |||
| AK139226.1 | - | 5 | 1897 | |||
| BC043077.1 | - | 5 | 1921 | 5721118 | ||
| BC046980.1 | - | 4 | 1741 | 6399438 | ||