Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LIMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204426.1 | NP_001191355.1 | 6 | 765 | ||
NM_002314.3 | NP_002305.1 | 7 | 987 | |||
GenBank mRNA | AB209557.1 | - | 4 | 459 | ||
AF134379.1 | - | 7 | 773 | |||
AK026544.1 | - | 1 | 23 | |||
AK290162.1 | - | 8 | 912 | |||
AK300382.1 | - | 6 | 704 | |||
D26309.1 | - | 8 | 868 |