Search
Search

Dctn5| Gene Symbol: Dctn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021608.3 | NP_067621.3 | 2, 3 | 214 | ||
| GenBank mRNA | AF190795.1 | - | 2, 3 | 185 | ||
| AK010238.1 | - | 2, 3 | 194 | |||
| AK051957.1 | - | 2, 3 | 153 | |||
| AK076100.1 | - | 2, 3 | 187 | |||
| AK076738.1 | - | 2, 3 | 214 | |||
| AK145958.1 | - | 1, 2 | 1165 | |||
| AK146123.1 | - | 2, 3 | 180 | |||
| AK151193.1 | - | 2, 3 | 186 | |||
| AK151876.1 | - | 2, 3 | 186 | |||
| AK168983.1 | - | 2, 3 | 194 | |||
| AK187397.1 | - | 2, 3 | 131 | |||
| BC005671.1 | - | 1, 2 | 1188 | 3153175 | ||
| BC010834.1 | - | 2, 3 | 183 | 4235312 | ||
| BC092273.1 | - | 1, 2 | 1167 | |||