Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam129a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022018.3 | NP_071301.2 | 10 | 1380 | ||
GenBank mRNA | AB049355.1 | - | 10 | 1220 | ||
AK029847.1 | - | 6 | 692 | |||
AK136597.1 | - | 10 | 1290 | |||
AK170193.1 | - | 10 | 1303 | |||
BC021332.1 | - | 10 | 1268 | |||
BC038006.1 | - | 10 | 1236 | |||
BC058234.1 | - | 8 | 980 | 6306849 | ||
BC099690.1 | - | 10 | 1287 | |||
BC137843.1 | - | 6 | 757 | 8860862 |