Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022020.2 | NP_071303.1 | 2 | 145 | ||
GenBank mRNA | AF260923.1 | - | 2 | 146 | ||
AY141971.1 | - | 2 | 155 | |||
BC028432.1 | - | 2 | 142 | 1348253 |