Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tnmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022322.2 | NP_071717.1 | 1, 2 | 257 | ||
GenBank mRNA | AB055422.1 | - | 1, 2 | 64 | ||
AF191768.1 | - | 1, 2 | 203 | |||
AF219993.1 | - | 1, 2 | 130 | |||
AF291655.1 | - | 1, 2 | 282 | |||
AK003748.1 | - | 1, 2 | 143 | |||
BC006919.1 | - | 1, 2 | 66 | 3601223 | ||
BC049944.1 | - | 1, 2 | 257 | 2811628 |