Search
Search

LMX1B| Gene Symbol: LMX1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174146.1 | NP_001167617.1 | 8 | 1104 | ||
| NM_001174147.1 | NP_001167618.1 | 8 | 1092 | |||
| NM_002316.3 | NP_002307.2 | 8 | 1071 | |||
| GenBank mRNA | AF057135.1 | - | 8 | 995 | ||
| BC069588.1 | - | 8 | 1011 | |||
| BC069601.1 | - | 8 | 995 | 7262383 | ||
| BC112120.1 | - | 8 | 1045 | 8327588 | ||
| BC113491.1 | - | 8 | 1045 | 8322543 | ||
| BC143801.1 | - | 8 | 1078 | 9052315 | ||
| KU178166.1 | - | 8 | 995 | |||
| KU178167.1 | - | 8 | 1016 | |||