Search Thermo Fisher Scientific
Gene Symbol: Klf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023184.3 | NP_075673.1 | 2 | 1051 | ||
GenBank mRNA | AF317225.1 | - | 1 | 926 | ||
AK007479.1 | - | 1 | 253 | |||
AK009739.1 | - | 2 | 1051 | |||
AK138824.1 | - | 2 | 1090 | |||
AK142618.1 | - | 2 | 1053 | |||
AK183052.1 | - | 1 | 32 | |||
AK205291.1 | - | 1 | 36 | |||
BC013486.1 | - | 1 | 971 | 4211067 |